Skip to Content
Merck
All Photos(1)

Key Documents

901221

Sigma-Aldrich

1-Fluoro-3-isopropoxybenzene

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11FO
CAS Number:
Molecular Weight:
154.18
MDL number:
UNSPSC Code:
12352200

Assay

≥95%

form

liquid

functional group

fluoro

InChI

1S/C9H11FO/c1-7(2)11-9-5-3-4-8(10)6-9/h3-7H,1-2H3

InChI key

INDGWZOOYLDIPO-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanistic Insight Leads to a Ligand Which Facilitates the Palladium?Catalyzed Formation of 2?(Hetero) Arylaminooxazoles and 4?(Hetero) Arylaminothiazoles.
Olsen E P, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 129(35), 10705-10708 (2017)
Sébastien Schmitt et al.
The Journal of organic chemistry, 78(3), 1222-1229 (2013-01-10)
Two concise and high-yielding diastereoselective syntheses of 4-aryl-3-methyl-4-piperidinemethanols were realized from 1,3-dimethyl-4-piperidinone. The key reactions to control the C3-C4 relative stereochemistry were the alkoxymethylation of a metalloenamine generated from 4-aryl-3-methyl-1,2,3,6-tetrahydropyridine that afforded the (3R*,4S*)-form and the nucleophilic substitution of a
Lukas Hintermann et al.
Organic letters, 10(21), 4859-4862 (2008-10-10)
A solvent-controlled inversion of leaving group ability allows selective access to either of two internal substitution products in S(N)Ar reactions of substrates with competing leaving groups. Application of this principle in a selective synthesis of the highly functionalized xanthone core
Solvent-controlled leaving-group selectivity in aromatic nucleophilic substitution.
Hintermann L, et al.
Organic Letters, 10(21), 4859-4862 (2008)
Esben P K Olsen et al.
Angewandte Chemie (International ed. in English), 56(35), 10569-10572 (2017-06-13)
By using mechanistic insight, a new ligand (EPhos) for the palladium-catalyzed C-N cross-coupling between primary amines and aryl halides has been developed. Employing an isopropoxy group at the C3-position favors the C-bound isomer of the ligand-supported palladium(II) complexes and leads

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service