Merck
  • Diastereoselective syntheses of (3R*,4R*)- and (3R*,4S*)-4-aryl-3-methyl-4-piperidinemethanol and fluoro analogues.
Diastereoselective syntheses of (3R*,4R*)- and (3R*,4S*)-4-aryl-3-methyl-4-piperidinemethanol and fluoro analogues.
The Journal of organic chemistry (2013-01-10)
Sébastien Schmitt, Richard C D Brown, Cécile Perrio
ABSTRACT
Two concise and high-yielding diastereoselective syntheses of 4-aryl-3-methyl-4-piperidinemethanols were realized from 1,3-dimethyl-4-piperidinone. The key reactions to control the C3-C4 relative stereochemistry were the alkoxymethylation of a metalloenamine generated from 4-aryl-3-methyl-1,2,3,6-tetrahydropyridine that afforded the (3R*,4S*)-form and the nucleophilic substitution of a fluoroarene with deprotonated 3-methyl-4-piperidinenitrile giving the (3R*,4R*)-isomer. The corresponding fluoromethyl analogues were subsequently obtained through the fluorination of the piperidinemethanols using DAST.