Skip to Content
Merck
All Photos(1)

Key Documents

901192

Sigma-Aldrich

1-Azido-2-(2-methoxyethoxy)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N3O2
CAS Number:
Molecular Weight:
145.16
MDL number:
UNSPSC Code:
12352202
Product 901192 is not currently sold in your country. Contact Technical Service

form

liquid

Quality Level

availability

available only in USA

storage temp.

2-8°C

SMILES string

[N+](=[N-])=NC(OC)COCC

InChI

1S/C5H11N3O2/c1-3-10-4-5(9-2)7-8-6/h5H,3-4H2,1-2H3

InChI key

PHMWJDNVPGHYAD-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service