Skip to Content
Merck
All Photos(2)

Key Documents

901106

Sigma-Aldrich

FIr6

Synonym(s):

Bis(2,4-difluorophenylpyridinato)-tetrakis(1-pyrazolyl)borate iridium(III)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$458.00

$458.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$458.00

About This Item

Empirical Formula (Hill Notation):
C34H24N10BF4Ir
CAS Number:
Molecular Weight:
851.64
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

$458.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

Quality Level

form

solid

color

yellow

SMILES string

FC(C=C1[Ir]23[N]4=CC=CN4B(N5N=CC=C5)(N6C=CC=N6)N7C=CC=[N]37)=CC(F)=C1C8=[N]2C=CC=C8

InChI

1S/C12H12BN8.C11H6F2N.Ir/c1-5-14-18(9-1)13(19-10-2-6-15-19,20-11-3-7-16-20)21-12-4-8-17-21;12-8-4-5-9(10(13)7-8)11-3-1-2-6-14-11;/h1-12H;1-4,6-7H;

InChI key

OXHBKGURKPVEIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improvement of efficiency roll-off in blue phosphorescence OLED using double dopants emissive layer.
Yoo S, et al.
Journal of Luminescence, 160, 346-350 (2015)
Yu Gu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(22), 8250-8256 (2015-04-23)
An adamantane-based host material, namely, 4-{3-[4-(9H-carbazol-9-yl)phenyl]adamantan-1-yl}benzonitrile (CzCN-Ad), was prepared by linking an electron-donating carbazole unit and an electron-accepting benzonitrile moiety through an adamantane bridge. In this approach, two functional groups were attached to tetrahedral points of adamantane to construct an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service