Skip to Content
Merck
All Photos(1)

Key Documents

901039

Sigma-Aldrich

mDCBPy

Synonym(s):

(3,5-Di(9H-carbazol-9-yl)phenyl)(pyridin-4-yl)methanone, mDCBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H23N3O
CAS Number:
Molecular Weight:
513.59
UNSPSC Code:
12352200

Assay

≥98%

Quality Level

form

solid

Orbital energy

HOMO 5.72 eV 
LUMO 2.72 eV 

General description

mDCBPy is a thermally activated delayed fluorescent (TADF) emitter, bearing a benzoylpyridine core as an electron accepting unit and two meta carbazolyl groups as electron donating units. This TADF emitter exhibits high-contrast reversible tunable emission and can be utilized in rewritable media. Blue and green OLEDs using mDCBP as the emitter achieved a maximum EQE value of 18.4 to 14.7%, and brightness reached up to 8900 cd m-2.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A thermally activated delayed blue fluorescent emitter with reversible externally tunable emission.
Rajamalli P, et al.
Journal of Material Chemistry C, 4, 900-904 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service