Skip to Content
Merck
All Photos(2)

Key Documents

D91535

Sigma-Aldrich

1,1′-Diethyl-4,4′-carbocyanine iodide

96%

Synonym(s):

Cryptocyanine, Kryptocyanine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$77.60

$77.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$77.60

About This Item

Empirical Formula (Hill Notation):
C25H25IN2
CAS Number:
Molecular Weight:
480.38
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$77.60


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

solid

mp

250.5-253 °C (lit.)

λmax

648 nm
703 nm (2nd)

SMILES string

[I-].CCN1C=CC(=C\C=C\c2cc[n+](CC)c3ccccc23)c4ccccc14

InChI

1S/C25H25N2.HI/c1-3-26-18-16-20(22-12-5-7-14-24(22)26)10-9-11-21-17-19-27(4-2)25-15-8-6-13-23(21)25;/h5-19H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

CEJANLKHJMMNQB-UHFFFAOYSA-M

Application

Suitable as laser dye

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wearable Organic Optoelectronic Sensors for Medicine.
Ashu K Bansal et al.
Advanced materials (Deerfield Beach, Fla.), 27(46), 7638-7644 (2014-12-10)
Benjamin Dietzek et al.
The journal of physical chemistry. B, 111(21), 6034-6041 (2007-05-08)
Applying optimal control to photoinduced trans-cis isomerization in condensed phase, the dynamics of bond-twisting motion of 1,1'-diethyl-4,4'-cyanine in methanol and propanol is revealed. The shape of the optimized pulse resulting from minimization of the photoisomer formation can be directly related
A Harriman et al.
Photochemistry and photobiology, 52(4), 735-740 (1990-10-01)
The photophysical properties of 1,1'-diethyl-4,4'-carbocyanine chloride (kryptocyanine) have been measured in methanol solution and for the dye bound to human serum albumin, incorporated in neutral micelles and after incubation with leukemia cells. In all cases, it is found that formation

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service