Skip to Content
Merck
All Photos(2)

Key Documents

900920

Sigma-Aldrich

Bis[[diisopropylphosphino]methyl]-4-methyl-pyridine bis(pivaloyloxy)cobalt

95%

Synonym(s):

Chirik cobalt borylation catalyst, JENNY cobalt borylation catalyst, JennyCat

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$347.00

$347.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$347.00

About This Item

Empirical Formula (Hill Notation):
C30H55CoNO4P2
CAS Number:
Molecular Weight:
614.64
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

$347.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

95%

form

powder

reaction suitability

core: cobalt
reagent type: catalyst

mp

>300 °C

SMILES string

CC1=CC(CP(C(C)C)C(C)C)=NC(CP(C(C)C)C(C)C)=C1.O=C(C(C)(C)C)O[Co]OC(C(C)(C)C)=O

InChI

1S/C20H37NP2.2C5H10O2.Co/c1-14(2)22(15(3)4)12-19-10-18(9)11-20(21-19)13-23(16(5)6)17(7)8;2*1-5(2,3)4(6)7;/h10-11,14-17H,12-13H2,1-9H3;2*1-3H3,(H,6,7);/q;;;+2/p-2

InChI key

JSODKCJQNNNNPE-UHFFFAOYSA-L

Application

Bis[[diisopropylphosphino]methyl]-4-methyl-pyridine bis(pivaloyloxy)cobalt also known as JennyCat is developed in the laboratory of Paul Chirik is an effective cobalt precatalyst for the borylation of aromatic compounds, including diverse heteroarenes, pyridine derivatives, and fluoroarenes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer V Obligacion et al.
Journal of the American Chemical Society, 139(7), 2825-2832 (2017-02-01)
Cobalt catalysts with electronically enhanced site selectivity have been developed, as evidenced by the high ortho-to-fluorine selectivity observed in the C(sp

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service