Skip to Content
Merck
All Photos(1)

Key Documents

900766

Sigma-Aldrich

6,12-Dihydro-6,6,12,12-tetrakis(4-octylphenyl)-2,8-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)dithieno[2,3-d:2′,3′-d′]-s-indaceno[1,2-b:5,6-b′]dithiophene

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$290.00

$290.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$290.00

About This Item

Empirical Formula (Hill Notation):
C88H112B2O4S4
CAS Number:
Molecular Weight:
1383.71
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

$290.00


Check Cart for Availability

Request a Bulk Order

Assay

>98%

Quality Level

form

solid

storage temp.

2-8°C

SMILES string

B%13(OC(C(O%13)(C)C)(C)C)c1[s]c2c([s]c3c2C(c6c3cc7c(c6)c8[s]c9c([s]c(c9)B%12OC(C(O%12)(C)C)(C)C)c8C7(c%11ccc(cc%11)CCCCCCCC)c%10ccc(cc%10)CCCCCCCC)(c5ccc(cc5)CCCCCCCC)c4ccc(cc4)CCCCCCCC)c1

InChI

1S/C88H112B2O4S4/c1-13-17-21-25-29-33-37-61-41-49-65(50-42-61)87(66-51-43-62(44-52-66)38-34-30-26-22-18-14-2)71-57-70-72(58-69(71)79-77(87)81-73(95-79)59-75(97-81)89-91-83(5,6)84(7,8)92-89)88(67-53-45-63(46-54-67)39-35-31-27-23-19-15-3,68-55-47-64(48-56-6

InChI key

QIKMIMQCQNEOFG-UHFFFAOYSA-N

Application

The product is an indacene monomer that can be used to prepare wide bandgap polymer, poly(6,6,12,12-tetrakis(4-hexylphenyl)-6,12-dihydro-dithieno[2,3-d:2′,3′-d′]-s-indaceno[1,2-b:5,6-b′]dithiophene) (PIDTT), for the development of polymer solar cells (PSCs) and organic thin film transistors (OTFTs).[1][2][3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?Benchtop? Biaryl Coupling Using Pd/Cu Cocatalysis: Application to the Synthesis of Conjugated Polymers
Minus, Matthew B, et al.
Organic Letters, 23(8), 2873-2877 (2021)
A wide band gap polymer based on indacenodithieno [3, 2-b] thiophene for high-performance bulk heterojunction polymer solar cells
Lee Woosung and Jung JW
Journal of Material Chemistry A, 5(2), 712-719 (2017)
Alkylated indacenodithieno [3, 2-b] thiophene-based all donor ladder-type conjugated polymers for organic thin film transistors
Lu Rimei, et al.
Journal of Material Chemistry C, 6(8), 2004-2009 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service