Skip to Content
Merck
All Photos(1)

Key Documents

900624

Sigma-Aldrich

Di-t-butyl oxaziridine

≥95%

Synonym(s):

Kurti oxaziridine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$315.00

$315.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$315.00

About This Item

Empirical Formula (Hill Notation):
C9H19NO
CAS Number:
Molecular Weight:
157.25
MDL number:
UNSPSC Code:
12352000
NACRES:
NA.22

$315.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

liquid

availability

available only in USA

refractive index

n/D 1.4453

density

0.90 g/mL

storage temp.

2-8°C

SMILES string

CC(C)(C)C1(NO1)C(C)(C)C

InChI

1S/C9H19NO/c1-7(2,3)9(10-11-9)8(4,5)6/h10H,1-6H3

InChI key

BEGIYSAPGHHUSW-UHFFFAOYSA-N

Application

This oxaziridine has been used in synthetic methods for amination, heteroatom transfer, and C-H functionalization. Recently, the lab of Laszlo Kurti demonstrated its application for direct electrophilic primary and secondary amination of arylmetals in the presence of Cu(I) salts -- void of precious metal catalysts, ligands, protecting groups, or directing groups.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F

Flash Point(C)

62 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Xu et al.
Nature chemistry, 7(10), 829-834 (2015-09-24)
Site-selective C-H functionalization has emerged as an attractive tool for derivatizing complex synthetic intermediates, but its use for late-stage diversification is limited by the functional groups that can be introduced, especially at unactivated sp(3)-hybridized positions. To overcome this, we introduce
Zhe Zhou et al.
Journal of the American Chemical Society, 139(1), 115-118 (2016-12-23)
Herein we disclose a novel method for the facile transfer of primary (-NH2) and secondary amino groups (-NHR) to heteroaryl- as well as arylcuprates at low temperature without the need for precious metal catalysts, ligands, excess reagents, protecting and/or directing
Hongyin Gao et al.
Nature chemistry, 9(7), 681-688 (2017-06-24)
Arylmetals are highly valuable carbon nucleophiles that are readily and inexpensively prepared from aryl halides or arenes and widely used on both laboratory and industrial scales to react directly with a wide range of electrophiles. Although C-C bond formation has
Synthesis of Alkoxylamines by Alkoxide Amination with 3,3'-Di-tert-butyloxaziridine.
Ingrid C. Choong et al.
The Journal of organic chemistry, 64(18), 6528-6529 (2001-10-25)

Related Content

Amines and their derivatives are ubiquitous substances since they make up the overwhelming majority of drug molecules, agrochemicals as well as many compounds that are produced by plants and living organisms (i.e., natural products).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service