Skip to Content
Merck
All Photos(1)

Key Documents

701742

Sigma-Aldrich

Copper(I) chloride - bis(lithium chloride) complex

1 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$276.00

$276.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$276.00

About This Item

Linear Formula:
CuCl · 2LiCl
Molecular Weight:
183.79
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$276.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

core: copper
reagent type: catalyst

concentration

1 M in THF

refractive index

n20/D 1.442

density

1.015 g/mL at 25 °C

SMILES string

[Li]Cl.[Li]Cl.Cl[Cu]

InChI

1S/3ClH.Cu.2Li/h3*1H;;;/q;;;3*+1/p-3

InChI key

QJRAZNXUNZEAQH-UHFFFAOYSA-K

Application

Solution used in Cu(I) transmetallation for amination reaction[1][2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F

Flash Point(C)

17.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kienle, M. et al.
Synthesis, 1272-1272 (2007)
General preparation of primary, secondary, and tertiary aryl amines by the oxidative coupling of polyfunctional aryl and heteroaryl amidocuprates.
Vicente Del Amo et al.
Angewandte Chemie (International ed. in English), 45(46), 7838-7842 (2006-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service