Skip to Content
Merck
All Photos(1)

Key Documents

900370

Sigma-Aldrich

(±)-Catechin-2,3,4-13C3 gallate

≥99 atom % 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,330.00

$1,330.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
$1,330.00

About This Item

Empirical Formula (Hill Notation):
13C3C19H18O10
Molecular Weight:
445.35
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.12

$1,330.00


Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Assay

≥97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

SMILES string

OC1=C(C(O)=CC(C(O[13C]([13C]2)[13C](OC3=C2C(O)=CC(O)=C3)C(C=C4O)=CC=C4O)=O)=C1)O

InChI

1S/C22H14O10/c23-11-6-14(25)12-8-19(32-22(30)10-4-16(27)20(29)17(28)5-10)21(31-18(12)7-11)9-1-2-13(24)15(26)3-9/h1-7,23-29H/i8+1,19+1,21+1

InChI key

SFJYIQKCXZHKAJ-SFWKTOTKSA-N

Related Categories

Application

Relative stereochemistry

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service