Skip to Content
Merck
All Photos(3)

Key Documents

22110

Sigma-Aldrich

(+)-Catechin hydrate

≥96.0% (sum of enantiomers, HPLC)

Synonym(s):

(+)-Cyanidol-3, (2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$61.80
10 G
$182.00
50 G
$707.00

$61.80


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$61.80
10 G
$182.00
50 G
$707.00

About This Item

Empirical Formula (Hill Notation):
C15H14O6 · xH2O
CAS Number:
Molecular Weight:
290.27 (anhydrous basis)
Beilstein:
3595244
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$61.80


Available to ship on16 April 2025Details


Request a Bulk Order

Assay

≥96.0% (sum of enantiomers, HPLC)

form

powder

optical activity

[α]/D +26±2°, c = 1 in H2O

impurities

≤8% water

mp

175-177 °C (anhydrous) (lit.)

storage temp.

2-8°C

SMILES string

[H]O[H].O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6.H2O/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7;/h1-5,13,15-20H,6H2;1H2/t13-,15+;/m0./s1

InChI key

OFUMQWOJBVNKLR-NQQJLSKUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Catechin hydrate is a polyphenolic flavonoid that exhibits antioxidant properties. It is commonly found in green tea, grape seeds, and bark of few trees like acacia and mahogany.[1]

Application

(+)-Catechin hydrate can be used:
  • As an inhibitor of steel corrosion in hydrochloric acid solution.[2]
  • As a model compound in the study of antimicrobial activities of flavonoids on Escherichia coli.[3]
  • As a starting material for the synthesis of catechin glucosides of biological importance.[4]

Caution

loses water of hydration during storage

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial mechanism of flavonoids against Escherichia coli ATCC 25922 by model membrane study
He M, et al.
Applied Surface Science, 305(5), 515-521 (2014)
Catechin hydrate inhibits proliferation and mediates apoptosis of SiHa human cervical cancer cells
Al-Hazzani AA and Alshatwi AA
Food And Chemical Toxicology, 49(12), 3281-3286 (2011)
Catechin glucosides: occurrence, synthesis, and stability
Raab T, et al.
Journal of Agricultural and Food Chemistry, 58(4), 2138-2149 (2010)
Electrochemical, thermodynamic and adsorption studies of (+)-catechin hydrate as natural mild steel corrosion inhibitor in 1 M HCl
Hussin MH and Kassim MJ
International Journal of Electrochemical Science, 6(5), 1396-1414 (2011)
Duckgue Lee et al.
International journal of molecular sciences, 20(24) (2019-12-19)
Endoplasmic reticulum (ER) stress is known to influence various cellular functions, including cell cycle progression. Although it is well known how ER stress inhibits cell cycle progression at the G1 phase, the molecular mechanism underlying how ER stress induces G2/M

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Protocols

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Questions

  1. How to dissolve (+)-catechin hydrate

    1 answer
    1. This product is tested for solubility at 50 mg/mL in hot ethanol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service