Skip to Content
Merck
All Photos(1)

Documents

852481

Sigma-Aldrich

6-Chloropurine riboside

99%

Synonym(s):

6-Chloropurine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClN4O4
CAS Number:
Molecular Weight:
286.67
Beilstein:
40573
EC Number:
MDL number:
UNSPSC Code:
12352123
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

158-162 °C (dec.) (lit.)

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(Cl)ncnc23

InChI

1S/C10H11ClN4O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2/t4-,6-,7-,10-/m1/s1

InChI key

XHRJGHCQQPETRH-KQYNXXCUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For a review of the geochemical and biological effects of this riboside, see Adv. Exp. Med. Biol. .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chem. Abstr., 106, 95594r-95594r (1987)
Cintia W Rivero et al.
Bioorganic & medicinal chemistry letters, 22(19), 6059-6062 (2012-09-11)
This work describes the application of thermophilic microorganisms for obtaining 6-halogenated purine nucleosides. Biosynthesis of 6-chloropurine-2'-deoxyriboside and 6-chloropurine riboside was achieved by Geobacillus stearothermophilus CECT 43 with a conversion of 90% and 68%, respectively. Furthermore, the selected microorganism was satisfactorily
Shigetada Kozai et al.
Nucleosides, nucleotides & nucleic acids, 22(5-8), 779-781 (2003-10-21)
A new method to introduce a benzyl group onto the 2'-OH of purine ribonucleoside is described. Thus, 6-chloropurine 3'-O-benzoylriboside and its 5'-O-trityl congener were condensed with benzyl alcohol using the Mitsunobu reaction to give the 2'-O-benzyl derivative. The yields were
Selection of purine nucleoside analogs based on multiple biological and biochemical parameters.
M J Noujaim et al.
Advances in experimental medicine and biology, 195 Pt B, 165-169 (1986-01-01)
J Alonso et al.
International journal of peptide and protein research, 34(1), 66-69 (1989-07-01)
The reaction of ribonuclease A with either 6-chloropurine riboside 5'-monophosphate or the corresponding nucleoside yields one derivative, with the reagent covalently bound to the alpha-amino group of Lys-1, called derivative II and derivative E, respectively. We studied by means of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service