Skip to Content
Merck
All Photos(3)

Key Documents

84190

Sigma-Aldrich

2-(2-Aminoethyl)-1,3-di-Boc-guanidine

≥90% (HPLC), for peptide synthesis

Synonym(s):

1,3-Di-Boc-2-(2-aminoethyl)guanidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$486.00
5 G
$1,380.00

$486.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$486.00
5 G
$1,380.00

About This Item

Empirical Formula (Hill Notation):
C13H26N4O4
CAS Number:
Molecular Weight:
302.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$486.00


Available to ship on16 April 2025Details


Request a Bulk Order

Product Name

2-(2-Aminoethyl)-1,3-di-Boc-guanidine, technical, ≥90% (HPLC)

grade

technical

Assay

≥90% (HPLC)

form

lumps

mp

95-98 °C

application(s)

peptide synthesis

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N\C(NC(=O)OC(C)(C)C)=N\CCN

InChI

1S/C13H26N4O4/c1-12(2,3)20-10(18)16-9(15-8-7-14)17-11(19)21-13(4,5)6/h7-8,14H2,1-6H3,(H2,15,16,17,18,19)

InChI key

GKQKBELHRNEGCI-UHFFFAOYSA-N

Other Notes

Guanidinylation reagent[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Nicolaou et al.
Bioorganic & medicinal chemistry, 6(8), 1185-1208 (1998-10-24)
Recent studies demonstrated that peptide and antibody antagonists of integrin alpha v beta 3 block angiogenesis and tumor growth. In this article, the design, synthesis and biological evaluation of a series of nitroaryl ether-based, nonpeptide mimetics are described. The design

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service