Skip to Content
Merck
All Photos(1)

Key Documents

275557

Sigma-Aldrich

1-Ethylguanidine sulfate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$92.10

$92.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$92.10

About This Item

Linear Formula:
[C2H5NHC(=NH)NH2]2·H2SO4
CAS Number:
Molecular Weight:
272.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

244 °C (dec.) (lit.)

functional group

amine

SMILES string

OS(O)(=O)=O.CCNC(N)=N.CCNC(N)=N

InChI

1S/2C3H9N3.H2O4S/c2*1-2-6-3(4)5;1-5(2,3)4/h2*2H2,1H3,(H4,4,5,6);(H2,1,2,3,4)

InChI key

UKQVDMIAGTYDFN-UHFFFAOYSA-N

Application

1-Ethylguanidine sulfate may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O G Busygina et al.
Biokhimiia (Moscow, Russia), 61(1), 119-125 (1996-01-01)
Guanidine thiol derivatives--a new class of soluble guanylate cyclase activators--have been studied. Guanidine thiols contain in their molecule both the guanidine and thiol groups which act as donors acceptors of nitric oxide (NO), respectively. The role of the guanidine thiol
J M Farley et al.
The Journal of physiology, 337, 753-768 (1983-04-01)
The effects of intracellular application of three compounds - QX314, N-ethylguanidine and N-octylguanidine - on the acetylcholine-activated ionic channels in chick myoballs were examined. Voltage-clamped myoballs were internally perfused using the suction-pipette technique. Changes in the internal ionic environment of
S M Vogel et al.
The Journal of general physiology, 83(6), 901-918 (1984-06-01)
Methyl- and ethylguanidine block the endplate current in frog muscle. Both derivatives blocked inward-going endplate currents without affecting outward endplate currents. Repetitive stimulation that evoked several inward endplate currents enhanced the block, which suggests that these agents interact with open

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service