Skip to Content
Merck
All Photos(1)

Key Documents

808024

Sigma-Aldrich

(S,R,R)-Elenor Amination Reagent

Synonym(s):

3H-3a,6-Methano-2,1-benzisothiazole-1(4H)-carboxamide, tetrahydro-N-hydroxy-8,8-dimethyl-, 2,2-dioxide, (3aS,6R,7aR)-

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$222.00

$222.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$222.00

About This Item

Empirical Formula (Hill Notation):
C11H18N2O4S
CAS Number:
Molecular Weight:
274.34
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

$222.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

O=C(N1S(C[C@]2(C3(C)C)CC[C@]3([H])C[C@@]12[H])(=O)=O)NO

InChI

1S/C11H18N2O4S/c1-10(2)7-3-4-11(10)6-18(16,17)13(8(11)5-7)9(14)12-15/h7-8,15H,3-6H2,1-2H3,(H,12,14)/t7-,8-,11-/m1/s1

InChI key

FYSORZDAOUCPGD-SOCHQFKDSA-N

Application

The following hydroxyamide is a chiral electrophilic nitroso reagent for the transfer of nitrogen to enol ethers. This methology was reported by the Read de Alaniz Group, is a mild approach that can be performed at room temperature and in the presence of air.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service