Skip to Content
Merck
All Photos(3)

Key Documents

8.18318

Sigma-Aldrich

D(-)-N-Methylglucamine

for resolution of racemates for synthesis

Synonym(s):

N-Methyl-D-glucamine, 1-Deoxy-1-methylaminosorbitol, 1-Deoxy-1-(methylamino)-D-glucitol, Meglumine

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$80.40

$80.40


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
250 G
$80.40

About This Item

Empirical Formula (Hill Notation):
C7H17NO5
CAS Number:
Molecular Weight:
195.21
Beilstein:
385906
MDL number:
UNSPSC Code:
12352104
EC Index Number:
228-506-9
NACRES:
NA.22

$80.40


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

form

solid

autoignition temp.

~662 °F

potency

>5000 mg/kg LD50, oral (Rat)

pH

11 (20 °C, 10 g/L in H2O)

bp

210 °C/1013 hPa (decomposes)

mp

129-131.5 °C (lit.)

solubility

1000 g/L

bulk density

300 kg/m3

storage temp.

2-30°C

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C7H17NO5/c1-8-2-4(10)6(12)7(13)5(11)3-9/h4-13H,2-3H2,1H3/t4-,5+,6+,7+/m0/s1

InChI key

MBBZMMPHUWSWHV-BDVNFPICSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 128 °C
Melting range (upper value): ≤ 131 °C
Spec. rotation [α²0/D (c=10 in water): -17.0 - -15.0 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service