Merck
CuI/DMPAO-catalyzed N-arylation of acyclic secondary amines.
Organic letters (2012-06-06)
Yu Zhang, Xinye Yang, Qizheng Yao, Dawei Ma
ABSTRACT
DMPAO has been found to be a powerful ligand to enable copper-catalyzed coupling of aryl halides with aliphatic acyclic secondary amines take place under relatively mild conditions, and coupling of aryl halides with primary amines and cyclic secondary amines proceeds at low catalyst loading.
Sigma-Aldrich
Copper(I) iodide, purum, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(I) iodide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Copper(I) iodide, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Dimethoxyanilino(oxo)acetic acid
Pricing and availability is not currently available.