Skip to Content
Merck
All Photos(1)

Key Documents

805742

Sigma-Aldrich

Sodium hexadecyl sulfate

contains ~40% sodium stearyl sulfate

Synonym(s):

Cetylsulfuric Acid Sodium Salt, Hexadecylsulfuric Acid Sodium Salt, Sodium Cetyl Sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$91.90

$91.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$91.90

About This Item

UNSPSC Code:
12352100
NACRES:
NA.22

$91.90


Check Cart for Availability

Request a Bulk Order

form

powder

mp

178 °C

Related Categories

General description

Sodium hexadecyl sulphate (SHS), a long chain sodium alkyl sulfate,[1] is a natural anionic surfactant.[2] Electrical conductivities of the aqueous solutions of SHS have been measured.[1] Mixtures of SHS and hexadecanol (HD) have been employed for the emulsion polymerization of styrene.[3] The presence of impurities in SHS can be evaluated by studying the impact of the chain length and pH of SHS on the electrokinetic properties of hematite.[4] SHS intercalated in hydrocalumite (a layer double hydroxide) has potential applications as sorbents for non-ionic organic contaminants.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emulsion polymerization of styrene with sodium hexadecyl sulphate/hexadecanol mixtures as emulsifiers. Initiation in monomer droplets.
Ugelstad J, et al.
Makromol. Chem., 175(2), 507-521 (1974)
Ping Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 151, 673-678 (2015-07-15)
Hydrocalumite (CaAl-LDH-Cl) interacted with a natural anionic surfactant, sodium hexadecyl sulfate (SHS), was performed using an intercalation method. To understand the intercalation behavior and characterize the resulting products, powder X-ray diffraction (XRD), scan electron microscopy (SEM) and mid-infrared (MIR) spectroscopy
The effect of sodium alkyl sulfates (C8-C16) on the electrokinetic properties of hematite.
Kosmulski M and Maczka E.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 492, 152-159 (2016)
The Electrical Conductivities of Aqueous Solutions of Sodium Dodecyl Sulphate and Sodium Hexadecyl Sulphate at Different Temperatures.
Howell OR and Robinson HGB.
Proceedings of the Royal Society of London A: Mathematical, Physical and Engineering Sciences., 155(885), 386-406 (1936)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service