Skip to Content
Merck
All Photos(1)

Key Documents

71443

Supelco

Sodium decyl sulfate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

Decyl sulfate sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$673.00

$673.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$673.00

About This Item

Linear Formula:
CH3(CH2)9OSO3Na
CAS Number:
Molecular Weight:
260.33
Beilstein:
3576860
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

$673.00


Estimated to ship on30 May 2025


Request a Bulk Order

description

anionic

Quality Level

Assay

≥99.0% (T)

form

flakes

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.05
λ: 220 nm Amax: 0.04
λ: 230 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Na+].CCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C10H22O4S.Na/c1-2-3-4-5-6-7-8-9-10-14-15(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1

InChI key

XZTJQQLJJCXOLP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium decyl sulfate is an anionic surfactant.[1] It is a sodium salt monodecyl ester of sulfuric acid. It also has application in textile, leather and other industries as emulsifier, wetting agent, dispersant, fiber lubricant, and synthetic fatliquor.[2]

Application

Sodium decyl sulfate was used in a study conducted to investigate the preferential interaction of poly(vinylpyrrolidone) (PVP) toward the alkyl sulfate surfactants using 13C and 1H NMR and NOESY measurements.[3]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. W. A. Milne
Gardner's Commercially Important Chemicals: Synonyms, Trade Names, and Properties, 560-560 (2005)
Binding of sodium decyl sulfate to a cationic polymer.
Shirahama, Keishiro, Hidefumi Yuasa, and Shinji Sugimoto.
Bulletin of the Chemical Society of Japan, 54.2, 375-377 (1981)
Complex formation between poly (vinylpyrrolidone) and sodium decyl sulfate studied through NMR.
Roscigno, Paola, et al.
Langmuir, 19.23, 9638-9644 (2003)
Miriam Gochin et al.
Journal of medicinal chemistry, 52(14), 4338-4344 (2009-06-19)
The hydrophobic pocket within the coiled coil domain of HIV-1 gp41 is considered to be a hot-spot suitable for small molecule intervention of fusion, although so far it has yielded only microM inhibitors. Previous peptide studies have identified specific hydrophobic
Francisco J Prado-Prado et al.
Bioorganic & medicinal chemistry, 18(6), 2225-2231 (2010-02-27)
There are many of pathogen parasite species with different susceptibility profile to antiparasitic drugs. Unfortunately, almost QSAR models predict the biological activity of drugs against only one parasite species. Consequently, predicting the probability with which a drug is active against

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service