Skip to Content
Merck
All Photos(2)

Key Documents

802999

Sigma-Aldrich

2-Acetamido-2-deoxy-β-D-glucopyranosylamine

97% (HPLC)

Synonym(s):

2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose, N-Acetyl-D-glucosylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$811.00

$811.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$811.00

About This Item

Empirical Formula (Hill Notation):
C8H16N2O5
CAS Number:
Molecular Weight:
220.22
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$811.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97% (HPLC)

form

powder

mp

60 °C

storage temp.

2-8°C

SMILES string

CC(=O)NC1C(N)OC(CO)C(O)C1O

InChI

1S/C8H16N2O5/c1-3(12)10-5-7(14)6(13)4(2-11)15-8(5)9/h4-8,11,13-14H,2,9H2,1H3,(H,10,12)

InChI key

MCGXOCXFFNKASF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This compound has been used as a building block in a variety of synthetic reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan Joseph et al.
Organic letters, 15(4), 732-735 (2013-01-26)
Herein is described the chemoselective Cu(II)-HOBt promoted chemical ligation of glycosylamines and peptide thioacids to give N-glycosylated peptides. The method is distinguished from other chemical approaches to peptide N-glycosylation in that (1) it can be employed in the presence of
Convergent solid-phase synthesis of N-glycopeptides facilitated by pseudoprolines at consensus-sequence Ser/Thr residues.
Vera Ullmann et al.
Angewandte Chemie (International ed. in English), 51(46), 11566-11570 (2012-09-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service