Skip to Content
Merck
All Photos(1)

Key Documents

670995

Sigma-Aldrich

2-Acetamido-2-deoxy-β-D-glucopyranosyl azide

≥98% (HPLC)

Synonym(s):

2-Acetamido-1-azido-1,2-dideoxy-β-D-glucopyranose

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$379.00

$379.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$379.00

About This Item

Empirical Formula (Hill Notation):
C8H14N4O5
CAS Number:
Molecular Weight:
246.22
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.22

$379.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

optical activity

[α]/D -24.5±1.5°, c = 1 in H2O

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1N=[N+]=[N-]

InChI

1S/C8H14N4O5/c1-3(14)10-5-7(16)6(15)4(2-13)17-8(5)11-12-9/h4-8,13,15-16H,2H2,1H3,(H,10,14)/t4-,5-,6-,7-,8-/m1/s1

InChI key

CCZCDCDASZJFGK-FMDGEEDCSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service