Skip to Content
Merck
All Photos(2)

Key Documents

802247

Sigma-Aldrich

2-Chloromethyl-2,3-dihydrothieno[3,4-b]-1,4-dioxine

95%

Synonym(s):

2-Chloromethyl EDOT, 2-Chloromethyl-2,3-dihydro-thieno[3,4-b][1,4]dioxine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$196.00

$196.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$196.00

About This Item

Empirical Formula (Hill Notation):
C7H7ClO2S
CAS Number:
Molecular Weight:
190.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$196.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

39-44 °C

transition temp

flash point >230 °F

functional group

chloro

SMILES string

ClCC1COc2cscc2O1

InChI

1S/C7H7ClO2S/c8-1-5-2-9-6-3-11-4-7(6)10-5/h3-5H,1-2H2

InChI key

UOXHKOVMJCKBOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloromethyl-2,3-dihydrothieno[3,4-b]-1,4-dioxine (EDOT-Cl) can be prepared by reacting 3,4-dimethoxythiophene with 3-chloro-1,2-propanediol via transesterification.[1] The six-memebered ring of the molecule exhibits twisted conformation.[2] It acts as an intermediate during the multi-step synthesis of poly(sulfobetaine-3,4-ethylenedioxythiophene) (PSBEDOT), a conjugated polymer.[3]

Application

2-Chloromethyl EDOT is a key building block for the incorporation of EDOT (3,4-ethylenedioxythio-phene) into copolymers.
It may be used in the synthesis of the following EDOT derivatives:[1]
  • 2-[((6-hexyloxy-9,10-anthraquinone)-2-yl)oxymethyl]-2,3-dihydrothieno[3,4-b][1,4]dioxine (AQ-EDOT)
  • 2-[(N′-(10-nonadecyl)perylene-3,4,9,10-biscarboximide-N-yl)4-phenoxymethyl]-2,3-dihydrothieno[3,4-b][1,4]dioxine (PTCDI-EDOT)
  • 2-[((6-hexyloxy-11,11,12,12-tetracyanoanthraquinodimethane)-2-yl)-oxymethyl]-2,3-dihydrothieno[3,4-b][1,4]dioxine (TCAQ-EDOT)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Chloromethyl-2,3-dihydrothieno [3,4-b][1,4] dioxine.
Xu J, et al.
Acta Crystallographica Section E, Structure Reports Online, 65(4), o668- o668 (2009)
Synthesis and electronic properties of anthraquinone-, tetracyanoanthraquinodimethane-, and perylenetetracarboxylic diimide-functionalized poly (3, 4-ethylenedioxythiophenes).
Segura JL, et al.
Chemistry of Materials, 18(12), 2834-2847 (2006)
Electroactive poly (sulfobetaine-3,4-ethylenedioxythiophene)(PSBEDOT) with controllable antifouling and antimicrobial properties.
Cao B, et al.
Chemical Science, 7(3), 1976-1981 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service