Skip to Content
Merck
All Photos(1)

Key Documents

669210

Sigma-Aldrich

3,4-(2′,2′-Diethylpropylene)dioxythiophene

97%

Synonym(s):

3,3-Diethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepin, EDOT analog

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$371.00

$371.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$371.00

About This Item

Empirical Formula (Hill Notation):
C11H16O2S
CAS Number:
Molecular Weight:
212.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$371.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.534

density

1.109 g/mL at 25 °C

SMILES string

CCC1(CC)COc2cscc2OC1

InChI

1S/C11H16O2S/c1-3-11(4-2)7-12-9-5-14-6-10(9)13-8-11/h5-6H,3-4,7-8H2,1-2H3

InChI key

HMKHKPKUPVRLOW-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Conjugated polymers offer charge transport between inorganic, electrically conducting metals and organic, proton-conducting biological systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service