Skip to Content
Merck
All Photos(2)

Key Documents

797308

Sigma-Aldrich

4,4′-Bis(1,2,2-triphenylvinyl)-1,1′-biphenyl

Synonym(s):

4,4′-Bis(1,2,2-triphenylethenyl)biphenyl, 4,4′-Bis(triphenylethenyl)-1,1′-biphenyl, BTPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H38
CAS Number:
Molecular Weight:
662.86
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98%

form

powder

SMILES string

C(C1=CC=CC=C1)(C2=CC=CC=C2)=C(C3=CC=CC=C3)C4=CC=C(C(C=C5)=CC=C5C(C6=CC=CC=C6)=C(C7=CC=CC=C7)C8=CC=CC=C8)C=C4

InChI

1S/C52H38/c1-7-19-41(20-8-1)49(42-21-9-2-10-22-42)51(45-27-15-5-16-28-45)47-35-31-39(32-36-47)40-33-37-48(38-34-40)52(46-29-17-6-18-30-46)50(43-23-11-3-12-24-43)44-25-13-4-14-26-44/h1-38H

InChI key

AYYZYPOAOFTMPC-UHFFFAOYSA-N

General description

4,4′-Bis(1,2,2-triphenylvinyl)-1,1′-biphenyl ( BTPE) is a luminogen. It is used to design BTPE-based electroluminescence devices.[1] Recently, it was used as a bluish-green emitter and host for organic lightemitting diodes (OLEDs). [2]

Application

4,4′-Bis(1,2,2-triphenylvinyl)biphenyl (BTPE) nanowires can be formed by annealing the crystalline microfibers. [2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth methods, enhanced photoluminescence, high hydrophobicity and light scattering of 4, 4?-bis (1, 2, 2-triphenylvinyl) biphenyl nanowires.
Chen S, et al.
Organic Electronics null
Zujin Zhao et al.
Chemical communications (Cambridge, England), 46(5), 686-688 (2010-01-21)
Two is better than one: a luminogen comprised of two units of tetraphenylethene (BTPE) emits more efficiently than that with one tetraphenylethene unit in the solid state; self-assembly of the BTPE molecules affords crystalline microfibers that fluoresce in 100% efficiency
Growth methods, enhanced photoluminescence, high hydrophobicity and light scattering of 4, 4?-bis (1, 2, 2-triphenylvinyl) biphenyl nanowires.
Chen S, et al.
Organic Electronics, 1996-2002 null
Aggregation-induced emission, self-assembly, and electroluminescence of 4, 4?-bis (1, 2, 2-triphenylvinyl) biphenyl.
Zhao Z, et al.
Chemical Communications (Cambridge, England), 686-688 null
Aggregation-induced emission, self-assembly, and electroluminescence of 4, 4?-bis (1, 2, 2-triphenylvinyl) biphenyl.
Zhao Z, et al.
Chemical Communications (Cambridge, England) null

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service