Skip to Content
Merck
All Photos(2)

Key Documents

797219

Sigma-Aldrich

4,4′-(1,2-Diphenylethene-1,2-diyl)diphenol

Synonym(s):

1,2-Bis(4-hydroxyphenyl)-1,2-diphenylethylene, 4,4′-(1,2-Diphenyl-1,2-ethenediyl)bisphenol, TPE-DOH

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$241.00

$241.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$241.00

About This Item

Empirical Formula (Hill Notation):
C26H20O2
CAS Number:
Molecular Weight:
364.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$241.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

Quality Level

form

powder

mp

225 °C (lit.)

SMILES string

OC(C=C1)=CC=C1/C(C2=CC=CC=C2)=C(C3=CC=CC=C3)/C4=CC=C(O)C=C4

InChI

1S/C26H20O2/c27-23-15-11-21(12-16-23)25(19-7-3-1-4-8-19)26(20-9-5-2-6-10-20)22-13-17-24(28)18-14-22/h1-18,27-28H/b26-25+

InChI key

ZYIGFXHZSKIVOO-OCEACIFDSA-N

Application

TPE-DOH is a synthetic intermediate of aggregation-induced emission (AIE) dye for use in further synthesis of alkyl-halogen to make ether and polymer reaction via esterification.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile synthesis of soluble nonlinear polymers with glycogen-like structures and functional properties from ?simple? acrylic monomers
Rongrong Hu; Jacky W. Y. Lam; Yong Yu; Herman H. Y. Sung; Ian D. Williams; et al
Polym. Chem., 4, 95-95 (2013)

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service