Skip to Content
Merck
All Photos(1)

Documents

777331

Sigma-Aldrich

Cy3-alkyne

for copper catalyzed click labeling

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73N5O10S3
Molecular Weight:
964.31
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

solid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

SMILES string

CC1(C)C(/C=C/C=C2N(CCCS([O-])(=O)=O)C3=CC=C(S(=O)([O-])=O)C=C3C/2(C)C)=[N+](CCCCCC(NCC#C)=O)C4=C1C=C(S(=O)([O-])=O)C=C4.CC[NH+](CC)CC.CC[NH+](CC)CC

InChI

1S/C35H43N3O10S3.2C6H15N/c1-6-19-36-33(39)14-8-7-9-20-37-29-17-15-25(50(43,44)45)23-27(29)34(2,3)31(37)12-10-13-32-35(4,5)28-24-26(51(46,47)48)16-18-30(28)38(32)21-11-22-49(40,41)42;2*1-4-7(5-2)6-3/h1,10,12-13,15-18,23-24H,7-9,11,14,19-22H2,2-5H3,(H3-,36,39,40,41,42,43,44,45,46,47,48);2*4-6H2,1-3H3

InChI key

OZKAPYXVMSRMDQ-UHFFFAOYSA-N

Application

Alkyne modified cyanine dye. Can be used to detect or label azide-containing molecules or biomolecules by fluorescence spectroscopy following azide-alkyne cycloaddition.

Solubility: DMSO, water, DMF, MeOH
Abs/Em: 553/566 nm
Extinction Coefficient: 151,000 cm-1M-1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jozef Mravec et al.
Scientific reports, 7(1), 15988-15988 (2017-11-24)
Auxin is a key plant regulatory molecule, which acts upon a plethora of cellular processes, including those related to cell differentiation and elongation. Despite the stunning progress in all disciplines of auxin research, the mechanisms of auxin-mediated rapid promotion of
Xiaoyu Li et al.
Molecular cell, 68(5), 993-1005 (2017-11-07)
Gene expression can be post-transcriptionally regulated via dynamic and reversible RNA modifications. N

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service