Skip to Content
Merck
All Photos(1)

Documents

777315

Sigma-Aldrich

Cy3-azide

90% (HPLC)

Synonym(s):

Cy3 Azide Fluroescent Probe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H44N6O10S3 · xC6H16N
Molecular Weight:
804.95 (salt-free basis)
UNSPSC Code:
12352125
NACRES:
NA.22

Quality Level

Assay

90% (HPLC)

form

solid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

Application

Azide modified cyanine dye. Can be used to detect or label alkyne- or cyclooctyne-containing molecules or biomolecules by fluorescence spectroscopy following azide-alkyne cycloaddition.
Solubility -DMSO; water; DMF; MeOH
Abs/Em - 553/566 nm
Extinction Coefficient - 151; 000 cm-1M-1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service