Skip to Content
Merck
All Photos(1)

Documents

774057

Sigma-Aldrich

Zirconium yttrium alloy

sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis (excluding <=1% Hf)

Synonym(s):

Zr/Y

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr.85Y.15
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

99.9% trace metals basis (excluding <=1% Hf)

form

solid

diam. × thickness

2.00 in. × 0.25 in.

Application

Solid Oxide Fuel cells operating at temperatures below 800 C (also known as intermediate temperature solid oxide fuel cell, IT-SOFC) are currently the topic of much research and development owing to the high degradation rates and materials costs incurred for SOFC operating at temperatures above 900 C. Thin films of electrode and electrolyte layers is one of the ways to achieve high performances in IT-SOFC. Zirconium yttrium alloy sputtering targets can be used to deposit thin films of YSZ layers with high electronic conductivities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hana, Sukreen;
Thin Solid Films, 517, 5830-5830 (2009)
Uhlenbruck, S.;
Solid State Ionics, 181, 447-447 (2010)
Lai, Bo-Kuai;
Journal of Power Sources, 196, 6299-6299 (2011)
Jee, Youngseok;
Journal of the Korean Ceramic Society, 47, 82-82 (2010)

Articles

This article describes the use and advantages of polyoxometalate-based redox-flow batteries as electrochemical energy storage systems over Li-ion batteries.

Learn about the different low temperature direct alcohol fuel cells based on nanostructured material catalysts and anion exchange membrane fuel cells.

Thin film solid oxide fuel cells offer technical advantages; review of TF-SOFC deposition methods for electrolyte thin films.

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service