Skip to Content
Merck
All Photos(1)

Documents

772585

Sigma-Aldrich

4-Amino-1-methylpyrazole

95%

Synonym(s):

1-Methyl-1H-pyrazol-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N3
CAS Number:
Molecular Weight:
97.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.556

bp

45-80 °C/760 mmHg

density

1.141 g/mL at 25 °C

storage temp.

−20°C

InChI

1S/C4H7N3/c1-7-3-4(5)2-6-7/h2-3H,5H2,1H3

InChI key

LBGSWBJURUFGLR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service