Skip to Content
Merck
All Photos(1)

Key Documents

700940

Sigma-Aldrich

4-Aminopyrazole

95%

Synonym(s):

1H-Pyrazol-4-ylamine, 1H-Pyrazole-4-amine, 4-Amino-1H-pyrazole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$186.00

$186.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$186.00

About This Item

Empirical Formula (Hill Notation):
C3H5N3
CAS Number:
Molecular Weight:
83.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$186.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder

mp

77-82 °C

SMILES string

Nc1cn[nH]c1

InChI

1S/C3H5N3/c4-3-1-5-6-2-3/h1-2H,4H2,(H,5,6)

InChI key

AXINVSXSGNSVLV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Beat J Brüschweiler et al.
Regulatory toxicology and pharmacology : RTP, 88, 214-226 (2017-06-29)
Azo dyes represent the by far most important class of textile dyes. Their biotransformation by various skin bacteria may release aromatic amines (AAs) which might be dermally absorbed to a major extent. Certain AAs are well known to have genotoxic
Bradley O Elmore et al.
PloS one, 10(5), e0125027-e0125027 (2015-05-06)
Serum lipoproteins (LP) are increasingly being recognized as dual purpose molecules that contribute to both cholesterol homeostasis and host innate defense. In fact, very low LP levels are associated with increased risk of bacterial infection in critically ill patients. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service