Skip to Content
Merck
All Photos(1)

Key Documents

763136

Sigma-Aldrich

Chloro(2-di-tert-butylphosphino-3,4,5,6-tetramethyl-2′,4′,6′-triisopropylbiphenyl)gold(I)

97%

Synonym(s):

Me4-tert-butyl XPhos-AuCl

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$191.00

$191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$191.00

About This Item

Empirical Formula (Hill Notation):
C33H53AuClP
CAS Number:
Molecular Weight:
713.17
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$191.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

core: gold
reagent type: catalyst

mp

221-224 °C (decomposition)

SMILES string

Cl[Au].CC(C)c1cc(C(C)C)c(c(c1)C(C)C)-c2c(C)c(C)c(C)c(C)c2P(C(C)(C)C)C(C)(C)C

InChI

1S/C33H53P.Au.ClH/c1-19(2)26-17-27(20(3)4)30(28(18-26)21(5)6)29-24(9)22(7)23(8)25(10)31(29)34(32(11,12)13)33(14,15)16;;/h17-21H,1-16H3;;1H/q;+1;/p-1

InChI key

IVIUDGKUDMXYRK-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service