Skip to Content
Merck
All Photos(1)

Documents

665177

Sigma-Aldrich

Chloro[tris(para-trifluoromethylphenyl)phosphine]gold(I)

99%

Synonym(s):

[Tris(para-trifluoromethylphenyl)phosphine]gold(I) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H12AuClF9P
CAS Number:
Molecular Weight:
698.70
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

reaction suitability

core: gold
reagent type: catalyst

SMILES string

Cl[Au].FC(F)(F)c1ccc(cc1)P(c2ccc(cc2)C(F)(F)F)c3ccc(cc3)C(F)(F)F

InChI

1S/C21H12F9P.Au.ClH/c22-19(23,24)13-1-7-16(8-2-13)31(17-9-3-14(4-10-17)20(25,26)27)18-11-5-15(6-12-18)21(28,29)30;;/h1-12H;;1H/q;+1;/p-1

InChI key

UNHMCLFZEQKHCQ-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We are proud to offer a treasure-trove of gold precatalysts and silver salts, as well as an extensive portfolio of unsaturated building blocks to accelerate your research success in this exciting field.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service