Skip to Content
Merck
All Photos(1)

Key Documents

753998

Sigma-Aldrich

PCDTBT

Synonym(s):

Poly[N-9′-heptadecanyl-2,7-carbazole-alt-5,5-(4′,7′-di-2-thienyl-2′,1′,3′-benzothiadiazole)], Poly[[9-(1-octylnonyl)-9H-carbazole-2,7-diyl]-2,5-thiophenediyl-2,1,3-benzothiadiazole-4,7-diyl-2,5-thiophenediyl]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C43H47N3S3)nC12H10
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Band gap: 1.9 eV

form

solid

mol wt

average Mw 100,000-140,000

loss

0.5 wt. % TGA, 409 °C

mp

270-300 °C

transition temp

Tm >400 °C

λmax

576 nm

Orbital energy

HOMO -5.5 eV 
LUMO -3.6 eV 

OPV Device Performance

ITO/MoO3-Al/PCDTBT:PC71BM/MoO3/Al

  • Short-circuit current density (Jsc): 10.88 mA/cm2
  • Open-circuit voltage (Voc): 0.88 V
  • Fill Factor (FF): 0.71
  • Power Conversion Efficiency (PCE): 6.77 %

ITO/PEDOT:PSS/PCDTBT:PC71BM (1:4)/TiOxAl
  • Short-circuit current density (Jsc): 11.32 mA/cm2
  • Open-circuit voltage (Voc): 0.88 V
  • Fill Factor (FF): 0.69
  • Power Conversion Efficiency (PCE): 6.9 %

ITO/PEDOT:PSS/PCDTBT:PC71BM/Al
  • Short-circuit current density (Jsc): 9.7 mA/cm2
  • Open-circuit voltage (Voc): 0.82 V
  • Fill Factor (FF): 0.61
  • Power Conversion Efficiency (PCE): 5.3 %

semiconductor properties

P-type (mobility=6×10−5 cm2/V·s)

Looking for similar products? Visit Product Comparison Guide

General description

PCDTBT is a carbozole based semiconducting co-polymer that is used as a donor material with a low band gap and a power efficiency of 9%. It has a quantum efficiency close to 100% that makes it a viable alternative of P3HT for a wide range of photovoltaics based applications.[1][2][3]
Soluble in THF, chloroform, chlorobenzene, dichlorobenzene, and 1,2,3-trichlorobenzene

Application

PCDTBT blend with PCBM as a nanocomposite can be used as a donor/acceptor material for the fabrication of photovoltaic solar cells and photovoltaic inks.[4][5][6] It may also be used as an active layer that can be used in the development of organic field effect transistors (OFETs) for the parts per million (ppm) level detection of NO2 gas.[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Aqueous PCDTBT: PC71BM Photovoltaic Inks Made by Nanoprecipitation
Prunet G, et al.
Macromolecular Rapid Communications, 39(2), 1700504-1700504 (2018)
Effect of traps on the charge transport in semiconducting polymer PCDTBT
Khan MT, et al.
Solid-State Electron, 145, 49-53 (2018)
Modeling of gate bias controlled NO2 response of the PCDTBT based organic field effect transistor
Kumar A, et al.
Chemical Physics Letters, 698(2), 7-10 (2018)
Ultrafast exciton dissociation followed by nongeminate charge recombination in PCDTBT: PCBM photovoltaic blends
Etzold F, et al.
Journal of the American Chemical Society, 133(24), 9469-9479 (2011)
Jung Hwa Seo et al.
Journal of the American Chemical Society, 133(22), 8416-8419 (2011-05-12)
The power conversion efficiencies of bulk heterojunction (BHJ) solar cells can be increased from 5 to 6.5% by incorporating an ultrathin conjugated polyelectrolyte (CPE) layer between the active layer and the metal cathode. Poly[N-9''-heptadecanyl-2,7-carbazole-alt-5,5-(4',7'-di-2-thienyl-2',1',3'-benzothiadiazole)] (PCDTBT) and [6,6]-phenyl C(71) butyric acid

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service