Skip to Content
Merck
All Photos(1)

Documents

743852

Sigma-Aldrich

COware PTFE discs

for COware, screwable two-chamber glass sytem

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352000
NACRES:
NA.22

form

disc

reaction suitability

reaction type: C-C Bond Formation

Citation

SyTracks CO-Generator – J. Am. Chem. Soc. 2011133, 6061-6071.
Carbonylative Mizoroki-Heck – Org. Lett. 201113, 2444-2447.
Primary Amides – Org. Lett. 201113, 4454-4457.
Protected Acids – Org. Lett. 2012, 14, 284-287.
N-Acylation of Ureas – J. Org. Chem. 201277, 3793-3799.
Carbonylative a-Arylation – Angew. Chem. Int. Ed. 201251, 798-801. VIP
Protected 1,3-Ketoaldehydes – Org. Lett. 201214, 2536-2539.
Alkoxy Carbonylation (ArCl) – J. Org. Chem. 2012, 77, 5357-5363.
Double Carbonylation – J. Org. Chem. 2012, 77, 6155-6165.
[14C]-Carbon Monoxide – J. Label. Compd. Radiopharm 201255, 411-418.

also commonly purchased with this product

replacement part


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

What is COgen? What is COware? What is the SyTracks COgenerator system? And more questions answered.

Related Content

The Skrydstrup group has developed reagents and glassware for carrying out transition metal catalyzed carbonylations in a simple and safe manner.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service