Skip to Content
Merck
All Photos(2)

Key Documents

8.43847

Sigma-Aldrich

2′-Methylacetoacetanilide

for synthesis

Synonym(s):

2′-Methylacetoacetanilide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$59.10

$59.10


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$59.10

About This Item

Empirical Formula (Hill Notation):
C11H13NO2
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
12352111
EC Index Number:
202-267-0
NACRES:
NA.22

$59.10


Estimated to ship on29 May 2025


Request a Bulk Order

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

Assay

≥98% (HPLC)

form

powder

potency

1600 mg/kg LD50, oral (Rat)

mp

103-106 °C

transition temp

flash point 143 °C

density

1.30 g/cm3

storage temp.

2-30°C

SMILES string

N(c1c(cccc1)C)C(=O)CC(=O)C

InChI

1S/C11H13NO2/c1-8-5-3-4-6-10(8)12-11(14)7-9(2)13/h3-6H,7H2,1-2H3,(H,12,14)

InChI key

TVZIWRMELPWPPR-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98 %
Melting range
- lower value:
≥ 103 °C
- upper value: ≤ 106 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service