Skip to Content
Merck
All Photos(1)

Key Documents

743615

Sigma-Aldrich

2-Methylpyridine

Arxada quality, ≥99.0% (GC)

Synonym(s):

2-Picoline, α-Picoline, 2-Methylpyridine, NSC 3409

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
Beilstein:
104581
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

vapor pressure

10 mmHg ( 24.4 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

995 °F

quality

Arxada quality

expl. lim.

8.6 %

manufacturer/tradename

Arxada AG

impurities

≤0.1% water

refractive index

n20/D 1.500 (lit.)

bp

128-129 °C (lit.)

mp

−70 °C (lit.)

density

0.942-0.946 at 20 °C
0.943 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccn1

InChI

1S/C6H7N/c1-6-4-2-3-5-7-6/h2-5H,1H3

InChI key

BSKHPKMHTQYZBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Nikolaos Kritikos et al.
Journal of chromatography. A, 1403, 70-80 (2015-06-06)
In the current study, quantitative structure-retention relationships (QSRR) were constructed based on data obtained by a LC-(ESI)-QTOF-MS/MS method for the determination of amino acid analogues, following their derivatization via chloroformate esters. Molecules were derivatized via n-propyl chloroformate/n-propanol mediated reaction. Derivatives
Shahul Hameed P et al.
Nature communications, 6, 6715-6715 (2015-04-01)
The widespread emergence of Plasmodium falciparum (Pf) strains resistant to frontline agents has fuelled the search for fast-acting agents with novel mechanism of action. Here, we report the discovery and optimization of novel antimalarial compounds, the triaminopyrimidines (TAPs), which emerged
Mir Ali Farajzadeh et al.
Biomedical chromatography : BMC, 29(12), 1921-1931 (2015-05-28)
In present study, a simultaneous derivatization and air-assisted liquid-liquid microextraction method combined with gas chromatography-nitrogen phosphorous detection has been developed for the determination of some phenolic compounds in biological samples. The analytes are derivatized and extracted simultaneously by a fast
Masazumi Tamura et al.
Nature communications, 6, 8580-8580 (2015-10-06)
Multidentate materials formed by simply mixing heterogeneous and homogeneous components are promising for construction of versatile active sites on the surface of heterogeneous compounds, however, to the best of our knowledge, there are no reports on such materials. Self-assembly of
Stéphanie Romand et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 63, 14-21 (2014-07-06)
A capillary isoelectric focusing (cIEF) method was developed for the determination of the ionization constants (pKa) of small molecules. Two approaches used to decrease the electroosmotic flow (EOF) were compared: (i) a hydroxypropylcellulose (HPC) coated capillary in aqueous medium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service