Merck
  • Triaminopyrimidine is a fast-killing and long-acting antimalarial clinical candidate.
Triaminopyrimidine is a fast-killing and long-acting antimalarial clinical candidate.
Nature communications (2015-04-01)
Shahul Hameed P, Suresh Solapure, Vikas Patil, Philipp P Henrich, Pamela A Magistrado, Sowmya Bharath, Kannan Murugan, Pavithra Viswanath, Jayashree Puttur, Abhishek Srivastava, Eknath Bellale, Vijender Panduga, Gajanan Shanbag, Disha Awasthy, Sudhir Landge, Sapna Morayya, Krishna Koushik, Ramanatha Saralaya, Anandkumar Raichurkar, Nikhil Rautela, Nilanjana Roy Choudhury, Anisha Ambady, Radha Nandishaiah, Jitendar Reddy, K R Prabhakar, Sreenivasaiah Menasinakai, Suresh Rudrapatna, Monalisa Chatterji, María Belén Jiménez-Díaz, María Santos Martínez, Laura María Sanz, Olivia Coburn-Flynn, David A Fidock, Amanda K Lukens, Dyann F Wirth, Balachandra Bandodkar, Kakoli Mukherjee, Robert E McLaughlin, David Waterson, Lyn Rosenbrier-Ribeiro, Kevin Hickling, V Balasubramanian, Peter Warner, Vinayak Hosagrahara, Adam Dudley, Pravin S Iyer, Shridhar Narayanan, Stefan Kavanagh, Vasan K Sambandamurthy
ABSTRACT
The widespread emergence of Plasmodium falciparum (Pf) strains resistant to frontline agents has fuelled the search for fast-acting agents with novel mechanism of action. Here, we report the discovery and optimization of novel antimalarial compounds, the triaminopyrimidines (TAPs), which emerged from a phenotypic screen against the blood stages of Pf. The clinical candidate (compound 12) is efficacious in a mouse model of Pf malaria with an ED99 <30 mg kg(-1) and displays good in vivo safety margins in guinea pigs and rats. With a predicted half-life of 36 h in humans, a single dose of 260 mg might be sufficient to maintain therapeutic blood concentration for 4-5 days. Whole-genome sequencing of resistant mutants implicates the vacuolar ATP synthase as a genetic determinant of resistance to TAPs. Our studies highlight the potential of TAPs for single-dose treatment of Pf malaria in combination with other agents in clinical development.
Product Description
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
Di-tert-butyl dicarbonate, ≥98.0% (GC)
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
Sigma-Aldrich
5-Bromouracil, 98%
Supelco
1,4-Dioxane, analytical standard
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
1,4-Dioxane, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)