Skip to Content
Merck
All Photos(1)

Key Documents

743453

Sigma-Aldrich

2-Aminoethyl sulfide ethyl Silica

≥45 μm particle size (95%)

Synonym(s):

PhosphonicS SEA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352309
NACRES:
NA.22

form

powder

extent of labeling

0.8-1.3 mmol/g loading

particle size

≥45 μm (95%)

Application

Silica Supported Metal and Organic Scavenger:
VO(II), Fe(III), Ni(II), Cu(II), Zn(II), Ru(III), Rh(I), Rh(III), Pd(0), Pd(II), Ag(I), Cd(II), Sn(II), Sn(IV), Os, Ir(I), Ir(III), Pt(II), Au(I), Au(III), Hg(II), Pb(II)

Legal Information

Sold in collaboration with PhosphonicS Ltd. for research purposes only.
PhosphonicS is a trademark of PhosphonicS Ltd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service