Skip to Content
Merck
All Photos(1)

Key Documents

745332

Sigma-Aldrich

Methyl thiourea ethyl sulfide ethyl Silica

≥45 μm particle size (95%)

Synonym(s):

PhosphonicS MTCf

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$308.00
50 G
$1,070.00

$308.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$308.00
50 G
$1,070.00

About This Item

UNSPSC Code:
12352309
NACRES:
NA.22

$308.00


Check Cart for Availability

Request a Bulk Order

form

powder

extent of labeling

0.8-1.4 mmol/g loading

particle size

≥45 μm (95%)

Application

Silica Supported Metal Scavenger:
Ru(III), Rh(I), Rh(III), Pd(0), Pd(II), Os, Ir(I), Ir(III), Pt(II)

Analysis Note

sulfur: detected

Legal Information

Sold in collaboration with PhosphonicS Ltd. for research purposes only.
PhosphonicS is a trademark of PhosphonicS Ltd.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service