Skip to Content
Merck
All Photos(1)

Key Documents

741299

Sigma-Aldrich

5-Bromo-2-(difluoromethyl)pyridine

95%

Synonym(s):

3-Bromo-6-(difluoromethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrF2N
CAS Number:
Molecular Weight:
208.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

35-40 °C

functional group

bromo
fluoro

storage temp.

2-8°C

SMILES string

FC(F)c1ccc(Br)cn1

InChI

1S/C6H4BrF2N/c7-4-1-2-5(6(8)9)10-3-4/h1-3,6H

InChI key

QXLZRIGSWWQOLG-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

192.9 °F

Flash Point(C)

89.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service