Skip to Content
Merck
All Photos(1)

Documents

734349

Sigma-Aldrich

Methyl 2-(hydroxymethyl)acrylate

contain inhibitor

Synonym(s):

Methyl 2-(hydroxymethyl)acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

contains

~1% MEHQ as inhibitor

refractive index

n20/D 1.456

density

1.129 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

COC(=O)C(=C)CO

InChI

1S/C5H8O3/c1-4(3-6)5(7)8-2/h6H,1,3H2,2H3

InChI key

RFUCOAQWQVDBEU-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.1 °F

Flash Point(C)

90.6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service