Skip to Content
Merck
All Photos(4)

Key Documents

212970

Sigma-Aldrich

2-(Bromomethyl)acrylic acid

98%

Synonym(s):

α-(Bromomethyl)acrylic acid, 2-(Bromomethyl)-2-propenoic acid, Bromomethylacrylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$129.00
10 G
$644.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$129.00
10 G
$644.00

About This Item

Linear Formula:
CH2=C(CH2Br)COOH
CAS Number:
Molecular Weight:
164.99
Beilstein:
2426204
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$129.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

70-73 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(=O)C(=C)CBr

InChI

1S/C4H5BrO2/c1-3(2-5)4(6)7/h1-2H2,(H,6,7)

InChI key

NOOYFQLPKUQDNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reacts with aldehydes and ketones to form α-methylene-butyrolactones.[1] Used in the synthesis of methotrexate analogues.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Abraham et al.
Journal of medicinal chemistry, 34(1), 222-227 (1991-01-01)
Five analogues of methotrextate (MTX), 10-deazaaminopterin (10-DAM), and 10-ethyl-10-deazaaminopterin (10-EDAM) in which the glutamate moiety was replaced by either a gamma-methyleneglutamate or beta-hydroxyglutamate were synthesized and evaluated for their antifolate activity. These analogous are 4-amino-4-deoxy-N10-methylpteroyl-beta-hydroxyglutamic acid (1), 4-amino-4-deoxy-10-deazapteroyl-beta-hydroxyglutamic acid (2)
Synthesis, 530-530 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service