Skip to Content
Merck
All Photos(1)

Key Documents

251208

Sigma-Aldrich

Ethyl 3-hydroxybenzoate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$61.60
100 G
$122.00

$61.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$61.60
100 G
$122.00

About This Item

Linear Formula:
HOC6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
Beilstein:
2208566
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$61.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

bp

187-188 °C/31 mmHg (lit.)

mp

71-73 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1cccc(O)c1

InChI

1S/C9H10O3/c1-2-12-9(11)7-4-3-5-8(10)6-7/h3-6,10H,2H2,1H3

InChI key

MWSMNBYIEBRXAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 3-hydroxybenzoate is a food-grade antimicrobial agent[1].

Application

Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Goretti et al.
International journal of food microbiology, 131(2-3), 178-182 (2009-03-10)
The in vitro antimycotic activity of a purified killer protein (KT4561) secreted by a strain of Williopsis saturnus was tested against 310 yeast strains belonging to 21 food spoilage species of 14 genera (Candida, Debaryomyces, Dekkera, Hanseniaspora, Issatchenkia, Kazachstania, Kluyveromyces
Mesomorphic properties of 1-(4'-dodecylbiphenyl-4-yl)-3-(2 or 3-alkoxyphenyl) propane-1, 3-diones: the influence of alkoxy-substituent position.
Sadashiva BK and Prasad V.
J. Chem. Soc. Perkin Trans. II, 4, 755-759 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service