Skip to Content
Merck
All Photos(1)

Key Documents

727016

Sigma-Aldrich

(R)-4-Fluoromandelic acid

ChiPros®, produced by BASF, 98%

Synonym(s):

4-Fluoro-D-mandelic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
8H7FO3
CAS Number:
Molecular Weight:
592.42
Beilstein:
2692850
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

produced by BASF

Assay

≥97.5% (HPLC)
98%

form

powder

optical purity

enantiomeric excess: ≥98.5%

functional group

carboxylic acid
fluoro
hydroxyl

SMILES string

O[C@@H](C(O)=O)C1=CC=C(F)C=C1

InChI

1S/C8H7FO3/c9-6-3-1-5(2-4-6)7(10)8(11)12/h1-4,7,10H,(H,11,12)/t7-/m1/s1

InChI key

RWCMOQXHIDWDDJ-SSDOTTSWSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service