Skip to Content
Merck
All Photos(1)

Key Documents

SML3664

Sigma-Aldrich

BTD

≥98% (HPLC)

Synonym(s):

6-​methyl-​N-​[3-​(tricyclo[3.3.1.13,​7]​dec-​1-​yloxy)​propyl]​-​4H-​1,​2,​4-​benzothiadiazine-​3-​propanamide 1,​1-​dioxide, N-[3-(adamantan-2-yloxy)propyl]-3-(6-methyl-1,1-dioxo-2H-1λ6,2,4-benzothiadiazin-3-yl)propanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$180.00
25 MG
$723.00

$180.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$180.00
25 MG
$723.00

About This Item

Empirical Formula (Hill Notation):
C24H33N3O4S
CAS Number:
Molecular Weight:
459.60
UNSPSC Code:
12352200
NACRES:
NA.77

$180.00


Estimated to ship on12 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(NCCCOC1(C2)C[C@H](C[C@H]2C3)C[C@H]3C1)CCC(NS4(=O)=O)=NC5=C4C=CC(C)=C5

Biochem/physiol Actions

BTD is a potent and selective activator of the transient receptor potential canonical channel 5 (TRPC5). BTD can also activate heteromeric channels composed of TRPC5 and its closest relatives TRPC1 or TRPC4.
Potent and selective TRPC5 activator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aisling Minard et al.
Cells, 7(6) (2018-06-06)
Proteins of the TRPC family can form many homo- and heterotetrameric cation channels permeable to Na⁺, K⁺ and Ca2+. In this review, we focus on channels formed by the isoforms TRPC1, TRPC4 and TRPC5. We review evidence for the formation
Holger Beckmann et al.
Cell calcium, 66, 10-18 (2017-08-16)
The transient receptor potential canonical channel 5 (TRPC5) is a Ca2+-permeable ion channel, which is predominantly expressed in the brain. TRPC5-deficient mice exhibit a reduced innate fear response and impaired motor control. In addition, outgrowth of hippocampal and cerebellar neurons

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service