Skip to Content
Merck
All Photos(1)

Documents

723088

Sigma-Aldrich

1-(11-Mercaptoundecyl)imidazole

96%

Synonym(s):

11-(1H-Imidazol-1-yl)undecane-1-thiol, MUI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26N2S
CAS Number:
Molecular Weight:
254.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

solid or liquid

mp

26-31 °C

storage temp.

2-8°C

SMILES string

SCCCCCCCCCCCn1ccnc1

InChI

1S/C14H26N2S/c17-13-9-7-5-3-1-2-4-6-8-11-16-12-10-15-14-16/h10,12,14,17H,1-9,11,13H2

InChI key

QBAITUUPZOBZFD-UHFFFAOYSA-N

General description

Binds a variety of metal ions onto a SAM. Porphyrins can be bound by axial ligand substitution as well as biological molecules such as cytochrome C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haniye Sadat Sajadi et al.
Cost effectiveness and resource allocation : C/E, 18, 20-20 (2020-07-03)
Building upon decades of continuous reforms, Iran has been implementing various initiatives to reach universal health coverage (UHC). Improving efficiency is a crucial intermediate policy objective for UHC. Therefore, this article aimed to measure the efficiency and productivity changes of the
J Springer et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 22(1), 80-86 (2015-09-25)
Invasive aspergillosis (IA) is associated with significant morbidity and mortality, and, among other factors, this is due to a delay in diagnosis performed with conventional techniques. A prospective, multicentre study was conducted to evaluate the efficacy of Aspergillus DNA screening
Lívia Oliveira Bezerra et al.
Games for health journal, 10(1), 43-49 (2020-07-28)
Objective: To verify whether pelvic floor muscle training (PFMT) associated with game therapy (GT) can potentiate improvements in PFM pressure, urinary loss, and perception of improvement in women with mixed urinary incontinence (MUI). Materials and Methods: A randomized and blinded
F Guo et al.
Journal of endocrinological investigation, 44(3), 443-452 (2020-06-10)
The use and contribution of prenatal multivitamins (PMV) as iodine source for pregnant women in China, especially in mildly iodine-deficient region, have not been well studied. This study aimed to explore the association between PMV intake during pregnancy and thyroid
Debjyoti Karmakar et al.
International urogynecology journal, 28(1), 95-100 (2016-07-06)
The Patient Global Impression of Improvement (PGI-I) and International Consultation of Incontinence Questionnaire - Short Form (ICIQ-SF) are validated instruments for the assessment of patient reported outcome measures (PROM) following treatment of stress urinary incontinence (SUI). However, there is a

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service