Skip to Content
Merck
All Photos(2)

Key Documents

720755

Sigma-Aldrich

Zinc di[bis(trifluoromethylsulfonyl)imide]

95%

Synonym(s):

Zn(NTf2)2

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$129.00

$129.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 G
$129.00

About This Item

Empirical Formula (Hill Notation):
C4F12N2O8S4Zn
CAS Number:
Molecular Weight:
625.68
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$129.00


In StockDetails


Request a Bulk Order

Quality Level

Assay

95%

form

solid

reaction suitability

core: zinc
reagent type: catalyst

mp

143-147 °C

SMILES string

FC(F)(F)S(=O)(=O)N([Zn]N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/2C2F6NO4S2.Zn/c2*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q2*-1;+2

InChI key

QEORIOGPVTWFMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Enantioselective Friedel-Crafts alkylation of pyrroles with β-CF3 acrylates
  • Stereoselective Diels-Alder reaction
  • Tandem cyclopropane ring-opening / Conia-ene cyclization
  • Aromatic nitration

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service