638021
DavePhos
97%
Synonym(s):
2-Dicyclohexylphosphino-2′-(N,N-dimethylamino)biphenyl
About This Item
Quality Level
Assay
97%
reaction suitability
reaction type: Cross Couplings
reagent type: ligand
reaction type: Arylations
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reagent type: ligand
reaction type: Hydroaminations
reagent type: ligand
reaction type: Kumada Coupling
reagent type: ligand
reaction type: Negishi Coupling
reagent type: ligand
reaction type: Suzuki-Miyaura Coupling
mp
121-124 °C (lit.)
functional group
phosphine
SMILES string
CN(C)c1ccccc1-c2ccccc2P(C3CCCCC3)C4CCCCC4
InChI
1S/C26H36NP/c1-27(2)25-19-11-9-17-23(25)24-18-10-12-20-26(24)28(21-13-5-3-6-14-21)22-15-7-4-8-16-22/h9-12,17-22H,3-8,13-16H2,1-2H3
InChI key
ZEMZPXWZVTUONV-UHFFFAOYSA-N
Application
- Palladium catalyzed sp3 arylation of 2-substituted-N-iminopyridinium ylides with different aryl chlorides to yield functionalized pyridines.
- Preparation of functionalized benzylic sulfones via palladium-catalyzed Negishi cross-coupling between alkyl sulfones and aryl halides.
- C-C Coupling of 3-haloquinolines with aryl sulfinates via palladium-catalyzed desulfitative arylation to form the corresponding biaryl compounds.
Learn more about Buchwald Phosphine Ligands
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service