Skip to Content
Merck
All Photos(1)

Key Documents

707120

SAFC

5-Chloro-7-iodoindole

96%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$242.00

$242.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest a Sample

Select a Size

Change View
250 MG
$242.00

About This Item

Empirical Formula (Hill Notation):
C8H5ClIN
CAS Number:
Molecular Weight:
277.49
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$242.00


Estimated to ship on25 May 2025


Request a Bulk OrderRequest a Sample

Assay

96%

form

solid

mp

77-82 °C

storage temp.

2-8°C

SMILES string

Clc1cc(I)c2[nH]ccc2c1

InChI

1S/C8H5ClIN/c9-6-3-5-1-2-11-8(5)7(10)4-6/h1-4,11H

InChI key

VRVJEJOGADKQRZ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service